Assay Details
| Gene Symbol: WTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590642.2 | XP_005590699.1 | 7 - 8 | 1103 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WTIP| Gene Symbol: WTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590642.2 | XP_005590699.1 | 7 - 8 | 1103 | 70 | |
Set Membership: |
|