Assay Details
| Gene Symbol: COPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563481.1 | XP_005563538.1 | 3 - 4 | 647 | 90 | |
| XM_005563482.2 | XP_005563539.1 | 2 - 3 | 489 | 90 | ||
| XM_005563483.2 | XP_005563540.1 | 2 - 3 | 400 | 90 | ||
| XM_015454930.1 | XP_015310416.1 | 2 - 3 | 710 | 90 | ||
| XM_015454932.1 | XP_015310418.1 | 3 - 4 | 445 | 90 | ||
| GenBank mRNA | AB070063.1 | - | 2 - 3 | 755 | 90 | |
More Information
Set Membership: |
|

COPS5
Genomic Map