Assay Details
| Gene Symbol: MTFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563434.2 | XP_005563491.1 | 2 - 3 | 245 | 94 | |
| XM_005563435.2 | XP_005563492.1 | 2 - 3 | 348 | 94 | ||
| XM_005563436.2 | XP_005563493.1 | 2 - 3 | 239 | 94 | ||
| XM_005563438.2 | XP_005563495.1 | 2 - 3 | 245 | 94 | ||
| XM_005563439.2 | XP_005563496.1 | 2 - 3 | 348 | 94 | ||
| XM_005563440.2 | XP_005563497.1 | 2 - 3 | 245 | 94 | ||
| XM_015454907.1 | XP_015310393.1 | 2 - 3 | 245 | 94 | ||
| GenBank mRNA | AB168272.1 | - | 2 - 3 | 234 | 94 | |
More Information
Set Membership: |
|

MTFR1
Genomic Map