Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285008.1 | NP_001271937.1 | 2 - 3 | 204 | 68 | |
| GenBank mRNA | AB174487.1 | - | 2 - 3 | 204 | 68 | |
More Information
Set Membership: |
|

SNRPD3
Genomic Map