Assay Details
| Gene Symbol: C1H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540223.2 | XP_005540280.1 | 1 - 2 | 391 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005540223.2 transcripts.
Set Membership: |
|

C1H1orf21
Genomic Map