Assay Details
| Gene Symbol: DNAJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284005.1 | NP_001270934.1 | 3 - 4 | 203 | 115 | |
| XM_015442699.1 | XP_015298185.1 | 2 - 3 | 332 | 115 | ||
| GenBank mRNA | AB174448.1 | - | 3 - 4 | 203 | 115 | |
More Information
Set Membership: |
|

DNAJA2
Genomic Map