Assay Details
Gene Symbol: SERGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578476.2 | XP_005578533.1 | 6 - 7 | 783 | 80 | |
XM_005578479.1 | XP_005578536.1 | 5 - 6 | 664 | 80 | ||
XM_015434995.1 | XP_015290481.1 | 6 - 7 | 786 | 80 | ||
XM_015434996.1 | XP_015290482.1 | 6 - 7 | 785 | 80 | ||
XM_015434997.1 | XP_015290483.1 | 6 - 7 | 783 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



