Assay Details
| Gene Symbol: NRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283490.1 | NP_001270419.1 | 4 - 5 | 570 | 93 | |
| XM_005542238.2 | XP_005542295.1 | 4 - 5 | 1178 | 93 | ||
| GenBank mRNA | AB220379.1 | - | 4 - 5 | 570 | 93 | |
More Information
Set Membership: |
|

NRAS
Genomic Map