Assay Details
| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567943.2 | XP_005568000.1 | 4 - 5 | 1450 | 77 | |
| XM_005567944.2 | XP_005568001.1 | 4 - 5 | 1450 | 77 | ||
| XM_015457414.1 | XP_015312900.1 | 4 - 5 | 3941 | 77 | ||
More Information
Set Membership: |
|

DGCR8
Genomic Map