Assay Details
| Gene Symbol: ELK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571917.1 | XP_005571974.1 | 3 - 4 | 1346 | 57 | |
| XM_015431413.1 | XP_015286899.1 | 3 - 4 | 1782 | 57 | ||
More Information
Set Membership: |
|

ELK3
Genomic Map