Assay Details
| Gene Symbol: SMARCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283128.1 | NP_001270057.1 | 990 | 63 | ||
| XM_005595361.2 | XP_005595418.1 | 1012 | 63 | |||
| GenBank mRNA | AB173747.1 | - | 990 | 63 | ||
More Information
Set Membership: |
|

SMARCB1
Genomic Map