Assay Details
| Gene Symbol: EFCAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563285.2 | XP_005563342.1 | 6 - 7 | 815 | 193 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFCAB1| Gene Symbol: EFCAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563285.2 | XP_005563342.1 | 6 - 7 | 815 | 193 | |
Set Membership: |
|