Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319371.1 | NP_001306300.1 | 8 - 9 | 856 | 93 | |
| XM_005577167.2 | XP_005577224.1 | 8 - 9 | 1634 | 93 | ||
| GenBank mRNA | AB173016.1 | - | 8 - 9 | 856 | 93 | |
More Information
Set Membership: |
|

BRMS1
Genomic Map