Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591016.1 | XP_005591073.1 | 4 - 5 | 1370 | 69 | |
| XM_015442657.1 | XP_015298143.1 | 4 - 5 | 1507 | 69 | ||
More Information
Set Membership: |
|

PKMYT1
Genomic Map