Assay Details
Gene Symbol: HADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283144.1 | NP_001270073.1 | 3 - 4 | 500 | 63 | |
GenBank mRNA | AB170801.1 | - | 3 - 4 | 500 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
