Assay Details
| Gene Symbol: EN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551259.1 | XP_005551316.1 | 1 - 2 | 903 | 76 | |
| GenBank mRNA | AY650377.1 | - | 245 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EN2| Gene Symbol: EN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551259.1 | XP_005551316.1 | 1 - 2 | 903 | 76 | |
| GenBank mRNA | AY650377.1 | - | 245 | 76 | ||
Set Membership: |
|