Assay Details
| Gene Symbol: CYP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589324.2 | XP_005589381.1 | 8 - 9 | 1692 | 74 | |
| XM_015441156.1 | XP_015296642.1 | 8 - 9 | 1147 | 74 | ||
More Information
Set Membership: |
|

CYP2S1
Genomic Map