Assay Details
| Gene Symbol: TSNAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284818.1 | NP_001271747.1 | 2 - 3 | 276 | 103 | |
| GenBank mRNA | AB169645.1 | - | 2 - 3 | 276 | 103 | |
More Information
Set Membership: |
|

TSNAX
Genomic Map