Assay Details
| Gene Symbol: PEX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284700.1 | NP_001271629.1 | 5 - 6 | 602 | 70 | |
| XM_005541260.2 | XP_005541317.1 | 4 - 5 | 511 | 70 | ||
| GenBank mRNA | AB170916.1 | - | 5 - 6 | 602 | 70 | |
More Information
Set Membership: |
|

PEX19
Genomic Map