Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283430.1 | NP_001270359.1 | 6 - 7 | 975 | 75 | |
| XM_015442114.1 | XP_015297600.1 | 6 - 7 | 1085 | 75 | ||
| GenBank mRNA | AB172210.1 | - | 6 - 7 | 975 | 75 | |
More Information
Set Membership: |
|

CD2BP2
Genomic Map