Assay Details
| Gene Symbol: GLP1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287663.1 | NP_001274592.1 | 3 - 4 | 289 | 68 | |
| GenBank mRNA | JN033215.1 | - | 3 - 4 | 289 | 68 | |
More Information
Set Membership: |
|

GLP1R
Genomic Map