Assay Details
| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284786.1 | NP_001271715.1 | 5 - 6 | 693 | 61 | |
| XM_015430659.1 | XP_015286145.1 | 5 - 6 | 903 | 61 | ||
| GenBank mRNA | AB174061.1 | - | 5 - 6 | 693 | 61 | |
More Information
Set Membership: |
|

PA2G4
Genomic Map