Assay Details
| Gene Symbol: GRIN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441472.1 | XP_015296958.1 | 2 - 3 | 902 | 75 | |
| XM_015441473.1 | XP_015296959.1 | 1 - 2 | 831 | 75 | ||
More Information
Set Membership: |
|

GRIN2D
Genomic Map