Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574014.2 | XP_005574071.1 | 3 - 4 | 1026 | 79 | |
| XM_005574016.2 | XP_005574073.1 | 3 - 4 | 2912 | 79 | ||
More Information
Set Membership: |
|

CTLA4
Genomic Map