Assay Details
| Gene Symbol: TRMT44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554381.2 | XP_005554438.1 | 1 - 2 | 657 | 111 | |
| XM_015450116.1 | XP_015305602.1 | 1 - 2 | 657 | 111 | ||
| XM_015450117.1 | XP_015305603.1 | 1 - 2 | 657 | 111 | ||
| XM_015450118.1 | XP_015305604.1 | 1 - 2 | 657 | 111 | ||
More Information
Set Membership: |
|

TRMT44
Genomic Map