Assay Details
| Gene Symbol: SENP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582768.2 | XP_005582825.1 | 6 - 7 | 1555 | 179 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SENP3| Gene Symbol: SENP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582768.2 | XP_005582825.1 | 6 - 7 | 1555 | 179 | |
Set Membership: |
|