Assay Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457243.1 | XP_015312729.1 | 5 - 6 | 1364 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXRED2| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457243.1 | XP_015312729.1 | 5 - 6 | 1364 | 150 | |
Set Membership: |
|