Assay Details
| Gene Symbol: RAB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540612.2 | XP_005540669.1 | 4 - 5 | 589 | 63 | |
| XM_005540613.2 | XP_005540670.1 | 4 - 5 | 608 | 63 | ||
| XM_005540614.2 | XP_005540671.1 | 4 - 5 | 578 | 63 | ||
| XM_005540615.2 | XP_005540672.1 | 4 - 5 | 604 | 63 | ||
| XM_005540617.2 | XP_005540674.1 | 3 - 4 | 272 | 63 | ||
| XM_015449013.1 | XP_015304499.1 | 4 - 5 | 2548 | 63 | ||
| XM_015449016.1 | XP_015304502.1 | 4 - 5 | 2530 | 63 | ||
| XM_015449019.1 | XP_015304505.1 | 4 - 5 | 2545 | 63 | ||
| XM_015449022.1 | XP_015304508.1 | 2 - 3 | 306 | 63 | ||
| XM_015449025.1 | XP_015304511.1 | 3 - 4 | 347 | 63 | ||
More Information
Set Membership: |
|

RAB29
Genomic Map