Assay Details
| Gene Symbol: SCG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284614.1 | NP_001271543.1 | 1 - 2 | 81 | 69 | |
| GenBank mRNA | AB169594.1 | - | 1 - 2 | 82 | 69 | |
| AB170865.1 | - | 1 - 2 | 82 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001284614.1 transcripts.
Set Membership: |
|

SCG2
Genomic Map