Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578315.1 | XP_005578372.1 | 5 - 6 | 647 | 117 | |
| XM_005578317.2 | XP_005578374.1 | 3 - 4 | 332 | 117 | ||
| XM_005578319.2 | XP_005578376.1 | 4 - 5 | 407 | 117 | ||
| XM_015434950.1 | XP_015290436.1 | 6 - 7 | 772 | 117 | ||
| XM_015434951.1 | XP_015290437.1 | 5 - 6 | 707 | 117 | ||
| XM_015434952.1 | XP_015290438.1 | 4 - 5 | 586 | 117 | ||
| XM_015434953.1 | XP_015290439.1 | 5 - 6 | 510 | 117 | ||
| XM_015434954.1 | XP_015290440.1 | 3 - 4 | 1926 | 117 | ||
| XM_015434955.1 | XP_015290441.1 | 3 - 4 | 1912 | 117 | ||
More Information
Set Membership: |
|

IMMP1L
Genomic Map