Assay Details
| Gene Symbol: EIF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562272.2 | XP_005562329.1 | 8 - 9 | 1897 | 96 | |
| XM_015454233.1 | XP_015309719.1 | 9 - 10 | 1587 | 96 | ||
| XM_015454234.1 | XP_015309720.1 | 9 - 10 | 1299 | 96 | ||
| GenBank mRNA | AB171209.1 | - | 8 - 9 | 1527 | 96 | |
More Information
Set Membership: |
|

EIF5
Genomic Map