Assay Details
| Gene Symbol: ACTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572970.2 | XP_005573027.1 | 2 - 3 | 444 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACTR3| Gene Symbol: ACTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572970.2 | XP_005573027.1 | 2 - 3 | 444 | 70 | |
Set Membership: |
|