Assay Details
| Gene Symbol: PSMC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283757.1 | NP_001270686.1 | 12 - 13 | 1448 | 102 | |
| XM_015437786.1 | XP_015293272.1 | 12 - 13 | 1284 | 102 | ||
| XM_015437787.1 | XP_015293273.1 | 11 - 12 | 1256 | 102 | ||
| XM_015437788.1 | XP_015293274.1 | 12 - 13 | 1294 | 102 | ||
| XM_015437789.1 | XP_015293275.1 | 12 - 13 | 1268 | 102 | ||
| GenBank mRNA | AB170233.1 | - | 11 - 12 | 1199 | 102 | |
| AB171188.1 | - | 11 - 12 | 1203 | 102 | ||
| AB171905.1 | - | 12 - 13 | 1448 | 102 | ||
More Information
Set Membership: |
|

PSMC5
Genomic Map