Assay Details
| Gene Symbol: HDGFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587572.2 | XP_005587629.1 | 4 - 5 | 623 | 62 | |
| XM_005587573.2 | XP_005587630.1 | 4 - 5 | 623 | 62 | ||
| XM_005587574.2 | XP_005587631.1 | 4 - 5 | 623 | 62 | ||
More Information
Set Membership: |
|

HDGFL2
Genomic Map