Assay Details
| Gene Symbol: CDHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447351.1 | XP_015302837.1 | 6 - 7 | 813 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDHR3| Gene Symbol: CDHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447351.1 | XP_015302837.1 | 6 - 7 | 813 | 84 | |
Set Membership: |
|