Assay Details
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452317.1 | XP_015307803.1 | 2 - 3 | 735 | 64 | |
XM_015452318.1 | XP_015307804.1 | 2 - 3 | 1385 | 64 | ||
XM_015452319.1 | XP_015307805.1 | 2 - 3 | 402 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
