Assay Details
| Gene Symbol: AIF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436320.1 | XP_015291806.1 | 2 - 3 | 329 | 92 | |
| XM_015436321.1 | XP_015291807.1 | 3 - 4 | 308 | 92 | ||
More Information
Set Membership: |
|

AIF1L
Genomic Map