Assay Details
| Gene Symbol: SRSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284068.1 | NP_001270997.1 | 3 - 4 | 493 | 62 | |
| XM_005576076.2 | XP_005576133.1 | 3 - 4 | 733 | 62 | ||
| XM_005576077.2 | XP_005576134.1 | 3 - 4 | 708 | 62 | ||
| XM_005576078.2 | XP_005576135.1 | 3 - 4 | 733 | 62 | ||
| GenBank mRNA | AB169747.1 | - | 3 - 4 | 496 | 62 | |
| AB170854.1 | - | 3 - 4 | 493 | 62 | ||
More Information
Set Membership: |
|

SRSF7
Genomic Map