Assay Details
| Gene Symbol: RPL3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590908.2 | XP_005590965.1 | 2 - 3 | 427 | 105 | |
| XM_005590909.2 | XP_005590966.1 | 2 - 3 | 296 | 105 | ||
More Information
Set Membership: |
|

RPL3L
Genomic Map