Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319623.1 | NP_001306552.1 | 9 - 10 | 1654 | 75 | |
| XM_005548300.2 | XP_005548357.1 | 8 - 9 | 1784 | 75 | ||
| GenBank mRNA | AB179448.1 | - | 9 - 10 | 1654 | 75 | |
More Information
Set Membership: |
|

TOMM70
Genomic Map