Assay Details
| Gene Symbol: P4HB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585264.2 | XP_005585321.1 | 1 - 2 | 366 | 55 | |
| GenBank mRNA | AB170168.1 | - | 1 - 2 | 205 | 55 | |
| AB170969.1 | - | 1 - 2 | 211 | 55 | ||
| AB171247.1 | - | 1 - 2 | 238 | 55 | ||
More Information
Set Membership: |
|

P4HB
Genomic Map