Assay Details
| Gene Symbol: BICD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570516.2 | XP_005570573.1 | 3 - 4 | 677 | 77 | |
| XM_005570518.2 | XP_005570575.1 | 3 - 4 | 580 | 77 | ||
| XM_005570519.2 | XP_005570576.1 | 3 - 4 | 754 | 77 | ||
| XM_005570521.2 | XP_005570578.1 | 3 - 4 | 580 | 77 | ||
| XM_005570522.2 | XP_005570579.1 | 3 - 4 | 580 | 77 | ||
More Information
Set Membership: |
|

BICD1
Genomic Map