Assay Details
| Gene Symbol: PPP4R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562036.2 | XP_005562093.1 | 2 - 3 | 307 | 114 | |
| XM_005562037.2 | XP_005562094.1 | 2 - 3 | 200 | 114 | ||
More Information
Set Membership: |
|

PPP4R3A
Genomic Map