Assay Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556125.2 | XP_005556182.1 | 3 - 4 | 849 | 76 | |
| XM_005556126.2 | XP_005556183.1 | 3 - 4 | 737 | 76 | ||
| XM_005556127.2 | XP_005556184.1 | 3 - 4 | 663 | 76 | ||
| XM_005556130.2 | XP_005556187.1 | 3 - 4 | 849 | 76 | ||
| XM_005556131.2 | XP_005556188.1 | 3 - 4 | 849 | 76 | ||
More Information
Set Membership: |
|

RBM46
Genomic Map