Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283806.1 | NP_001270735.1 | 8 - 9 | 835 | 76 | |
| GenBank mRNA | AF144132.1 | - | 8 - 9 | 836 | 76 | |
| X99971.1 | - | 835 | 76 | |||
More Information
Set Membership: |
|

DAZL
Genomic Map