Assay Details
| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564007.2 | XP_005564064.1 | 2 - 3 | 659 | 66 | |
| XM_005564008.2 | XP_005564065.1 | 2 - 3 | 974 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005564007.2, XM_005564008.2 transcripts.
Set Membership: |
|

ZHX2
Genomic Map