Assay Details
| Gene Symbol: TENT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556575.2 | XP_005556632.1 | 5 - 6 | 511 | 90 | |
| XM_015451202.1 | XP_015306688.1 | 4 - 5 | 1873 | 90 | ||
| XM_015451203.1 | XP_015306689.1 | 4 - 5 | 1874 | 90 | ||
| XM_015451204.1 | XP_015306690.1 | 4 - 5 | 1875 | 90 | ||
| XM_015451205.1 | XP_015306691.1 | 4 - 5 | 1873 | 90 | ||
More Information
Set Membership: |
|

TENT4A
Genomic Map