Assay Details
| Gene Symbol: SLC25A37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562848.2 | XP_005562905.1 | 2 - 3 | 643 | 68 | |
| XM_015454586.1 | XP_015310072.1 | 3 - 4 | 637 | 68 | ||
More Information
Set Membership: |
|

SLC25A37
Genomic Map