Assay Details
| Gene Symbol: SRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552939.2 | XP_005552996.1 | 2 - 3 | 1141 | 75 | |
| XM_015449386.1 | XP_015304872.1 | 2 - 3 | 2156 | 75 | ||
More Information
Set Membership: |
|

SRF
Genomic Map