Assay Details
| Gene Symbol: UPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549618.2 | XP_005549675.1 | 5 - 6 | 811 | 111 | |
| XM_005549619.2 | XP_005549676.1 | 5 - 6 | 671 | 111 | ||
More Information
Set Membership: |
|

UPP1
Genomic Map